Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP-D Antibody (119), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB333099 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333099 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333099 Supplier Novus Biologicals Supplier No. NBP290480APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SP-D Monoclonal antibody specifically detects SP-D in Mouse samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SP-D
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 119
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse SP-D (Accession#: NP_033186.1, Met1-Phe374)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6441
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.