Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SP-D Antibody Pair [HRP]
SDP

Catalog No. NB010133 Shop All R&D Systems Products
Click to view available options
Quantity:
15 Plates

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Mouse SP-D.

Specifications

Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse SP-D.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Gene ID (Entrez) 6441
Gene Symbol SFTPD
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-SP-D polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of rabbit anti-Mouse SP-D monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 105 ng of recombinant Mouse SP-D.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cell Cycle and Replication
    Storage Requirements Storage is content dependent.
    Target SP-D
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.6 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.