Learn More
Bel-Art™ Large Pails
Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Description
- Available in yellow or translucent white for easier viewing of fluid level
- Polypropylene is steam autoclavable (121°C), Polyethylene is not autoclavable
- 14-qt./13.2L pail has steel-plated swinging bail handle with grip
- Large pails meet US Specification MIL-P36257A
Compliance
Meet US Specification MIL-P36257A (8/10/70).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | Pail |
| Autoclavable | Not Autoclavable |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.