Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Transpette™ Eight Channel Transfer Pipettor
SCI_ED

Catalog No. S42970 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Sterility:
Sterile
Material:
Polyethylene
Quantity:
100/Cs.
Packaging Type:
Individually Wrapped

Perform plate washing, media change and supernatant collection without the use of pipettors and pipette tips with this inexpensive, eight-channel transfer pipettor.

  • 8-Channel with 600μL capacity per channel
  • Made of translucent plastic
  • Discard after hot radioactive uses to avoid radiation contamination of expensive liquid handling devices
  • Non-autoclavable
  • Available with sterile and non-sterile options

Specifications

Material Polyethylene
Sterility Sterile
For Use With (Application) For washing plates, feeding, and supernatant aspiration from microwell plates
Quantity 100/Cs.
Packaging Type Individually Wrapped
Color Natural
Volume (Metric) 600 μL
Length (Metric) 180 mm
Type Transfer Pipet
Disposable Single-use
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.