Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB600232549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600232549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600232549 Supplier Novus Biologicals Supplier No. NB600232JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SP1 Polyclonal specifically detects SP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SP1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Sp1 transcription factor, specificity protein 1, transcription factor Sp1, TSFP1
Gene Symbols SP1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 700 and 740 using the numbering given in entry XP_028606.4 (GeneID 6667).
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6667
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.