Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB600233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600233 Supplier Novus Biologicals Supplier No. NB600233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

SP1 Polyclonal specifically detects SP1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation Sequencing.

Specifications

Antigen SP1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP Assay
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunohistochemistry 1:2000 - 1:10000, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:2000 - 1:10000, Chromatin Immunoprecipitation (ChIP) 4 ug/30 ug lysate, Chromatin Immunoprecipitation Sequencing
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. P08047
Gene Alias Sp1 transcription factor, specificity protein 1, transcription factor Sp1, TSFP1
Gene Symbols SP1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 750 and the C-terminus (residue 785) using the numbering given in entry XP_028606.4 (GeneID 6667).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6667
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Chicken, Goat, Bovine, Dog, Horse, Rabbit, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant and Northern whitecheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.