Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP100 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB331917 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331917 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331917 Supplier Novus Biologicals Supplier No. NBP297407APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SP100 Polyclonal antibody specifically detects SP100 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SP100
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZp686E07254, FLJ00340, FLJ34579, lysp100b, nuclear antigen Sp100, nuclear autoantigen Sp-100, Nuclear dot-associated Sp100 protein, SP100 nuclear antigen, SP100-HMG nuclear autoantigen, Speckled 100 kDa
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SP100 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6672
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.