Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP100 Mouse anti-Human, Clone: PCRP-SP100-1B9, Novus Biologicals™
SDP

Catalog No. NB177516 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177516 100 μg
NB177517 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177516 Supplier Novus Biologicals Supplier No. NBP313780100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SP100 Monoclonal antibody specifically detects SP100 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen SP100
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone PCRP-SP100-1B9
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 0.5-2 ug/million cells, ELISA, Immunofluorescence 0.5-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P23497
Gene Alias DKFZp686E07254, FLJ00340, FLJ34579, lysp100b, nuclear antigen Sp100, nuclear autoantigen Sp-100, Nuclear dot-associated Sp100 protein, SP100 nuclear antigen, SP100-HMG nuclear autoantigen, Speckled 100 kDa
Host Species Mouse
Immunogen Recombinant full-length human SP100 protein (Uniprot: P23497)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6672
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.