Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP110 Antibody (OTI4C1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB274306A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274306A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274306A48 Supplier Novus Biologicals Supplier No. NBP274306AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SP110 Monoclonal specifically detects SP110 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SP110
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4C1
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ22835, IFI41, IFI75, interferon-induced protein 41, 30kD, Interferon-induced protein 41/75, interferon-induced protein 75, 52kD, IPR1, phosphoprotein 41, phosphoprotein 75, SP110 nuclear body protein, Speckled 110 kDa, Transcriptional coactivator Sp110, VODI
Gene Symbols SP110
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 232-521 of human SP110 (NP_004501) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3431.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.