Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPAG1 Antibody (10G1/2), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250301UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250301UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250301UV Supplier Novus Biologicals Supplier No. NBP250301UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPAG1 Monoclonal specifically detects SPAG1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SPAG1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 10G1/2
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ32920, HSD-3.8sperm-associated antigen 1, Infertility-related sperm protein Spag-1, SP75, sperm associated antigen 1, tetratricopeptide repeat-containing protein, TPIS, TPR-containing protein involved in spermatogenesis
Gene Symbols SPAG1
Host Species Mouse
Immunogen Synthetic peptide SPA-3 (sequence designed to avoid TPR motifs - KTAPFNKEKERRKIEIQEVNE)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 6674
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.