Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPAG16 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB329694 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329694 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329694 Supplier Novus Biologicals Supplier No. NBP297844APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPAG16 Polyclonal antibody specifically detects SPAG16 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SPAG16
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias DKFZp666P1710, FLJ22724, FLJ37717, Pf20 protein homolog, PF20MGC87036, sperm associated antigen 16, sperm-associated antigen 16 protein, sperm-associated WD repeat protein, WD repeat domain 29, WDR29
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SPAG16 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 79582
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.