Learn More
Description
Specifications
Specifications
| Antigen | SPAG9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.28mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal |
| Gene Alias | Cancer/testis antigen 89, c-Jun NH2-terminal kinase-associated leucine zipper protein, C-Jun-amino-terminal kinase-interacting protein 4, CT89JIP4, FLJ14006, FLJ26141, FLJ34602, HLC4, HLC-6, HSS, Human lung cancer oncogene 6 protein, JLPPHETSYD1FLJ13450, JNK interacting protein, JNK/SAPK-associated protein, JNK-associated leucine-zipper protein, JNK-interacting protein 4, KIAA0516JIP-4, lung cancer oncogene 4, MAPK8IP4, Max-binding protein, MGC117291, MGC14967, MGC74461, Mitogen-activated protein kinase 8-interacting protein 4, PIG6, proliferation-inducing gene 6, Proliferation-inducing protein 6, Protein highly expressed in testis, sperm associated antigen 9, Sperm surface protein, Sperm-associated antigen 9, Sperm-specific protein, Sunday driver 1 |
| Gene Symbols | SPAG9 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
