Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPANXN3 Antibody (OTI4B1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP274308R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274308R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274308R Supplier Novus Biologicals Supplier No. NBP274308R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPANXN3 Monoclonal specifically detects SPANXN3 in Human samples. It is validated for Western Blot.

Specifications

Antigen SPANXN3
Applications Western Blot
Classification Monoclonal
Clone OTI4B1
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cancer/testis antigen family 11, member 8, CT11.8, Nuclear-associated protein SPAN-Xn3, SPANX family member N3, SPANX family, member N3, SPANX-N3sperm protein associated with the nucleus on the X chromosome N3
Gene Symbols SPANXN3
Host Species Mouse
Immunogen Full length recombinant protein of human SPANXN3 (NP_001009609) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 139067.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.