Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPARC Antibody (122511), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB345719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345719 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345719 Supplier Novus Biologicals Supplier No. FAB941MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPARC Monoclonal specifically detects SPARC in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry (Intracellular Staining), Immunohistochemistry.

Specifications

Antigen SPARC
Applications CyTOF, ELISA, Flow Cytometry (Intracellular Staining), Immunohistochemistry
Classification Monoclonal
Clone 122511
Conjugate mFluor Violet 610 SE
Dilution ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin)
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SPARC, Ala18-Ile303, Accession # P09486
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6678
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.