Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPARC Antibody (124413), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB174525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174525 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB174525 Supplier Novus Biologicals Supplier No. FAB942J
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SPARC Monoclonal antibody specifically detects SPARC in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry

Specifications

Antigen SPARC
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 124413
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin)
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SPARC/Osteonectin, Ala18-Ile302, Accession # P07214
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6678
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.