Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Spare Molecular Model Parts
Shop All Eisco Products

Click to view available options
Product Type:
Ball with 1 Hole
Ball with 2 Holes
Ball with 2 Holes at 180i
Ball with 2 Holes at 90i
Ball with 3 Holes
Ball with 4 Holes
Ball with 5 Holes
Color:
Black
Blue
Purple
Red
White
Description
This 10 pack provides extras of this specific part for addition to a full set or to replace lost parts
- Part of a Molecular Model kit
- Pack of 10 Balls
Specifications
Specifications
Color | Blue |
For Use With (Application) | Molecular modeling |
Height (Metric) | 2.2 cm |
Includes | 10 balls |
Length (Metric) | 2.2 cm |
Material | Plastic |
Quantity | 10 |
Width (Metric) | 2.2 cm |
Product Type | Ball with 2 Holes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction