Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spastin Antibody (Sp 3G11-1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB385108 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385108 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385108 Supplier Novus Biologicals Supplier No. NBP308681R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Spastin Monoclonal antibody specifically detects Spastin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Spastin
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Sp 3G11-1
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias ADPSPSpastic paraplegia 4 protein, FSP2EC 3.6.4.3, KIAA1083EC 3.6.1.3, spastic paraplegia 4 (autosomal dominant; spastin), spastin, SPG4
Host Species Mouse
Immunogen Recombinant full-length human Spastin protein. (Uniprot: Q9UP0)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Cycle and Replication, Cellular Signaling, Epigenetics, metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6683
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.