Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA17 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1553720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1553720 20 μL
NBP155371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1553720 Supplier Novus Biologicals Supplier No. NBP15537120UL

Rabbit Polyclonal Antibody

SPATA17 Polyclonal specifically detects SPATA17 in Human samples. It is validated for Western Blot.

Specifications

Antigen SPATA17
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96L03
Gene Alias IQ motif containing H, IQCH, MSRG11, MSRG-11, RP11-144C20.1, spermatogenesis associated 17, spermatogenesis-associated protein 17, spermatogenesis-related protein 11
Gene Symbols SPATA17
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SPATA17(spermatogenesis associated 17) The peptide sequence was selected from the C terminal of SPATA17. Peptide sequence NMFLPFSSYHKNEKYIPSMHLSSKYGPISYKEQFRSENPKKWICDKDFQT.
Molecular Weight of Antigen 43 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128153
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.