Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA2 Antibody (Z1P1F10*F12), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250304UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250304UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250304UV Supplier Novus Biologicals Supplier No. NBP250304UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPATA2 Monoclonal specifically detects SPATA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen SPATA2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z1P1F10*F12
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias KIAA0757FLJ13167, PD1spermatogenesis associated PD1, spermatogenesis associated 2, spermatogenesis-associated protein 2, Spermatogenesis-associated protein PD1, tamo
Gene Symbols SPATA2
Host Species Mouse
Immunogen Peptide Sequence - KSTQLSHLVYR
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9825
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.