Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA20 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB047873 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047873 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047873 Supplier Novus Biologicals Supplier No. NBP297798AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA20 Polyclonal specifically detects SPATA20 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SPATA20
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp686H1839, FLJ21347, FLJ21969, MGC111032, sperm protein SSP411, spermatogenesis associated 20, spermatogenesis-associated protein 20, Sperm-specific protein 411, Ssp411, Tisp78, transcript increased in spermiogenesis 78
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SPATA20 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64847
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.