Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA22 Antibody (OTI6H9), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N274309J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274309J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274309J646 Supplier Novus Biologicals Supplier No. NBP274309JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPATA22 Monoclonal specifically detects SPATA22 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SPATA22
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6H9
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias NYDSP20, NYD-SP20, spermatogenesis associated 22, spermatogenesis-associated protein 22, testes development-related NYD-SP20, Testis development protein NYD-SP20
Gene Symbols SPATA22
Host Species Mouse
Immunogen Full length recombinant protein of human SPATA22 (NP_115987) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84690.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.