Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA25 Antibody (Z37P3H11*B2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP250305C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250305C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250305C Supplier Novus Biologicals Supplier No. NBP250305C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPATA25 Monoclonal specifically detects SPATA25 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen SPATA25
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z37P3H11*B2
Conjugate DyLight 650
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C20orf165, Chromosome 20 Open Reading Frame 165, DJ337O18.8, Spermatogenesis Associated 25, Testis-Specific Gene 23 Protein, TSG23
Gene Symbols SPATA25
Host Species Mouse
Immunogen Peptide Sequence - LVRSKRGQP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 128497
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.