Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA2L Antibody (OTI3F6), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP274310C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274310C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274310C Supplier Novus Biologicals Supplier No. NBP274310C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPATA2L Monoclonal specifically detects SPATA2L in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen SPATA2L
Applications Western Blot
Classification Monoclonal
Clone OTI3F6
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C16orf76, chromosome 16 open reading frame 76, MGC26885, SPATA2-like protein, spermatogenesis associated 2-like, spermatogenesis-associated protein 2-like protein, tamo
Gene Symbols SPATA2L
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 179-424 of human SPATA2L (NP_689552) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 124044.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.