Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA2L Antibody (Z2P2D5*C8), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP250306AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250306AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250306AI Supplier Novus Biologicals Supplier No. NBP250306AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPATA2L Monoclonal specifically detects SPATA2L in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SPATA2L
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Z2P2D5*C8
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C16orf76, chromosome 16 open reading frame 76, MGC26885, SPATA2-like protein, spermatogenesis associated 2-like, spermatogenesis-associated protein 2-like protein, tamo
Gene Symbols SPATA2L
Host Species Mouse
Immunogen Peptide Sequence - LLYNSPGARP
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124044
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.