Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB342280 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342280 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342280 Supplier Novus Biologicals Supplier No. NBP281816PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA3 Polyclonal antibody specifically detects SPATA3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SPATA3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias spermatogenesis associated 3, spermatogenesis-associated protein 3, testis and spermatogenesis cell apoptosis related gene 1, Testis spermatocyte apoptosis-related protein 1, TSARG1Testis and spermatogenesis cell-related protein 1
Host Species Rabbit
Immunogen SPATA3 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SPATA3. The immunogen is located within the last 50 amino acids of SPATA3. Amino Acid Squence: LTFYRKSSRKPSSHRNA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 130560
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.