Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA31A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB124321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB124321 100 μg
NB124322 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB124321 Supplier Novus Biologicals Supplier No. NBP309710100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA31A1 Polyclonal specifically detects SPATA31A1 in Human samples. It is validated for Western Blot.

Specifications

Antigen SPATA31A1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of Human SPATA31A1 (NP_001035154). Peptide sequence SSTPWVLDIFLTLVFALGFFFLLLPYLSYFRCDDPPSPSPGKRKCPVGRR
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 647060
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.