Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167152 20 μg
NB167151 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167152 Supplier Novus Biologicals Supplier No. NBP31602820UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPATA5 Polyclonal antibody specifically detects SPATA5 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen SPATA5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias AFG2ATPase family gene 2 homolog, ATPase family protein 2 homolog, SPAFspermatogenesis associated factor SPAF, spermatogenesis associated 5, Spermatogenesis-associated factor protein, spermatogenesis-associated protein 5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 228-304 of human SPATA5 (NP_001304728).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166378
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.