Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPATA6L Antibody, Novus Biologicals™
SDP

Catalog No. NBP215655 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215655 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215655 Supplier Novus Biologicals Supplier No. NBP215655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPATA6L Polyclonal specifically detects SPATA6L in Human samples. It is validated for Western Blot.

Specifications

Antigen C9orf68
Applications Western Blot
Classification Polyclonal
Concentration 0.67mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Alias bA6J24.2, C9orf68, chromosome 9 open reading frame 68, FLJ10058, hypothetical protein LOC55064, RP11-280I16.2, spermatogenesis associated 6-like
Gene Symbols SPATA6L
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human C9orf68. The exact sequence is proprietary.
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55064
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.