Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPC24 Antibody, Novus Biologicals™
SDP

Catalog No. NBP232105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232105 Supplier Novus Biologicals Supplier No. NBP232105
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPC24 Polyclonal specifically detects SPC24 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative).

Specifications

Antigen SPC24
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_872319.1
Gene Alias FLJ90806, hSpc24, kinetochore protein Spc24, SPBC24, SPC24, NDC80 kinetochore complex component, homolog (S. cerevisiae), spindle pole body component 24 homolog, spindle pole body component 24 homolog (S. cerevisiae)
Gene Symbols SPC24
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Spindle Pole Body Component 24 using the numbering given in entry NP_872319.1 (GeneID 147841).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147841
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.