Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPCS2 Antibody (OTI1E4), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP274312A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274312A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274312A70 Supplier Novus Biologicals Supplier No. NBP274312AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPCS2 Monoclonal specifically detects SPCS2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen SPCS2
Applications Western Blot
Classification Monoclonal
Clone OTI1E4
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias MGC117366, signal peptidase complex subunit 2, signal peptidase complex subunit 2 homolog (S. cerevisiae), SPC25
Gene Symbols SPCS2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-64 of human SPCS2 (NP_055567) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9789
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.