Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPDYE1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB031514 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB031514 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB031514 Supplier Novus Biologicals Supplier No. NBP2835850.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPDYE1 Polyclonal specifically detects SPDYE1 in Human samples. It is validated for Western Blot.

Specifications

Antigen SPDYE1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias Putative WBSCR19-Like Protein 6, Ringo1, SPDYB2L2, SPDYE, Speedy E, Speedy Homolog E1, Speedy Homolog E1 (Xenopus Laevis), Speedy Protein E1, Speedy/RINGO Cell Cycle Regulator Family Member E1, WBSCR19, Williams Beuren Syndrome Chromosome Region 19 Protein, Williams-Beuren Syndrome Chromosomal Region 19 Protein
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of Human SPDYE1
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285955
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.