Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPECC1L Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB051192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB051192 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB051192 Supplier Novus Biologicals Supplier No. NBP298024B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPECC1L Polyclonal specifically detects SPECC1L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SPECC1L
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Cytokinesis And Spindle Organization A, Cytospin A, CYTSA, GBBB2, KIAA0376, OBLFC1, Renal Carcinoma Antigen NY-REN-22, SPECC1-Like, SPECC1-Like Protein, Sperm Antigen With Calponin Homology And Coiled-Coil Domains 1-Like
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SPECC1L fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23384
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.