Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectinomycin DihydroChloride Pentahydrate, Cell Culture Grade, PROMO, Thermo Scientific Chemicals

Catalog No. J6182014PM
Click to view available options
Quantity:
25 g

Spectinomycin dihydrochloride pentahydrate is the hydrochloride salt form of spectinomycin, an aminocyclitol compound showing antibacterial and antimicrobial activities.

General Description

  • Spectinomycin dihydrochloride pentahydrate is derived from Streptomyces spectabilis and presents antibacterial and antimicrobial activities
  • Spectinomycin hydrochloride is a compound that inhibits the 30S ribosomal subunit of bacteria and interrupts protein synthesis and peptide elongation, leading to bacterial cell death

Application

  • Spectinomycin dihydrochloride pentahydrate is a compound that is active against gram-negative bacteria
  • In the laboratory, this compound is used in cell culture applications
  • It prevents contamination and can be used to select for resistant bacteria and plant cells containing the marker gene Spcr
  • It has been used to study respiratory tract infections caused by Pasteurella multocida and Mannheimia haemolytica
  • In genetic approaches, this compound can be used to amplify the plasmid copy number
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22189-32-8
Molecular Formula C14H36Cl2N2O12
Molecular Weight (g/mol) 495.34 g/mol
MDL Number MFCD00150886
InChI Key DCHJOVNPPSBWHK-UXXUFHFZSA-N
Synonym Spectinomycin dihydrochloride
PubChem CID 67108398
ChEBI CHEBI:9217
IUPAC Name dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.03,8]tetradecan-7-one pentahydrate dichloride
SMILES [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

Specifications

Type Spectinomycin Dihydrochloride Pentahydrate
Physical Form Powder
Color White
Quantity 25 g
Solubility Information Soluble in water, methanol, dimethyl sulfoxide, ethanol and dimethyl formamide.
Grade Cell Culture
Chemical Name or Material Spectinomycin dihydrochloride pentahydrate
EINECSNumber 244-554-3
RTECSNumber WG7400000
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.