Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin alpha 1 Antibody (SPTA1/2939R), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB387477 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387477 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387477 Supplier Novus Biologicals Supplier No. NBP308346DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Spectrin alpha 1 Monoclonal antibody specifically detects Spectrin alpha 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Spectrin alpha 1
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPTA1/2939R
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2)
Host Species Rabbit
Immunogen Recombinant fragment of human Spectrin alpha 1 (around aa356-475) (exact sequence is proprietary) (Uniprot: P02549)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 6708
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.