Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrin alpha 1 Antibody (SR1447), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322179100UL
Description
Spectrin alpha 1 Monoclonal antibody specifically detects Spectrin alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
Spectrin alpha 1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1447 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2) | |
A synthesized peptide derived from human Spectrin alpha 1 (Uniprot #: P02549) | |
100 μg | |
Cell Biology, Cellular Markers | |
6708 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction