Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin alpha 1 Antibody (SR1447), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$309.50 - $423.00

Specifications

Antigen Spectrin alpha 1
Clone SR1447
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB258937
SDP
View Documents
Novus Biologicals
NBP322179100UL
100 μg
Each for $423.00
Only null left
Add to Cart
 
NB258949
SDP
View Documents
Novus Biologicals
NBP32217950UL
50 μg
Each for $309.50
Only null left
Add to Cart
 
Description

Description

Spectrin alpha 1 Monoclonal antibody specifically detects Spectrin alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications

Specifications

Spectrin alpha 1
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2)
A synthesized peptide derived from human Spectrin alpha 1 (Uniprot #: P02549)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1447
Western Blot, Flow Cytometry, Immunofluorescence
Unconjugated
Rabbit
Cell Biology, Cellular Markers
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
6708
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.