Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin beta 1 Antibody (RG/26), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB386967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386967 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386967 Supplier Novus Biologicals Supplier No. NBP308440FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Spectrin beta 1 Monoclonal antibody specifically detects Spectrin beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Spectrin beta 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RG/26
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias beta-I spectrin, EL3, erythrocyte, HS2, HSPTB1, membrane cytoskeletal protein, spectrin, beta, erythrocytic, SPTB1
Host Species Mouse
Immunogen Red cell membranes
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 6710
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.