Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin beta 2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10040829 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10040829 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10040829 Supplier Novus Biologicals Supplier No. NB10040829
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Spectrin beta 2 Polyclonal specifically detects Spectrin beta 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Spectrin beta 2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q01082
Gene Alias beta-fodrin, beta-G spectrin, Beta-II spectrin, beta-spectrin 2, beta-spectrin II, betaSpII, ELF, embryonic liver beta-fodrin, Fodrin beta chain, spectrin beta chain, brain 1, spectrin, beta, non-erythrocytic 1, Spectrin, non-erythroid beta chain 1, SPTB2
Gene Symbols SPTBN1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2150 and 2200 of human Spectrin Non-Erythroid Beta 1 using the numbering given in entry NP_003119.2 (GeneID 6711).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6711
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque, European domestic ferret and Northern white-cheeked gibbon .
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.