Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin beta 3 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB339233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339233 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339233 Supplier Novus Biologicals Supplier No. NB11058346PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Spectrin beta 3 Polyclonal antibody specifically detects Spectrin beta 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Spectrin beta 3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Beta-III spectrin, glutamate transporter EAAT4-associated protein 41, GTRAP41, KIAA0302, SCA5, spectrin beta chain, brain 2, spectrin, beta, non-erythrocytic 2, Spectrin, non-erythroid beta chain 2, spinocerebellar ataxia 5
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a C-terminal region between residue 2340 and 2390 of Spectrin beta 3 using the numbering given in entry NP_008877.1
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6712
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.