Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin beta 3 Antibody (SPTBN2/1584), Novus Biologicals™
SDP

Catalog No. NB25331520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB25331520 20 μg
NB253315100 100 μg
NB253315200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB25331520 Supplier Novus Biologicals Supplier No. NBP25331520UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Spectrin beta 3 Monoclonal specifically detects Spectrin beta 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen Spectrin beta 3
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
Classification Monoclonal
Clone SPTBN2/1584
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias Beta-III spectrin, glutamate transporter EAAT4-associated protein 41, GTRAP41, KIAA0302, SCA5, spectrin beta chain, brain 2, spectrin, beta, non-erythrocytic 2, Spectrin, non-erythroid beta chain 2, spinocerebellar ataxia 5
Gene Symbols SPTBN2
Host Species Mouse
Immunogen Recombinant human SPTBN2 fragment (aa356-475) (exact sequence is proprietary)
Molecular Weight of Antigen 246 kDa
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6712
Target Species Human, Mouse, Rat
Content And Storage Store at 4C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.