Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrin beta-4 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$220.00 - $536.00
Specifications
| Antigen | Spectrin beta-4 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIPA5121581
|
Invitrogen™
PA5121581 |
20 μL |
Each for $220.00
|
|
|||||
PIPA5121580
![]() |
Invitrogen™
PA5121580 |
100 μL |
Each for $536.00
|
|
|||||
Description
Spectrin is the major constituent of the cytoskeletal network underlying the erthrocyte plasma membrane and determines cell shape, arrangement of transmembrane proteins and organization of organelles. It is expressed at very low levels in many tissues, with the strongest expression in the cerebellum, spinal cord, stomach, pituitary gland, liver, pancreas, salivary gland, kidney, bladder and heart. Spectrin beta 4 is a non-erythrocyte member, and is expressed in the brain and pancreatic islets and localisted to the nuclear matrix, cytoplasmic vesicles and PML nuclear bodies. Beta4 spectrins are also essential for membrane stability and the molecular organization of the nodes of Ranvier.Specifications
| Spectrin beta-4 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Pig | |
| Q9H254 | |
| 57731 | |
| Lys64-Ser285 (Accession Q9H254), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SPTBN4 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 1700022P15Rik; 5830426A08Rik; beta-IV spectrin; beta-spectrin 4; dyn; EMBL BAB83243.1; KIAA1642; lnd; lumbosacral neuroaxonal dystrophy; neuroaxonal dystrophy; nmf261; nmf379; quivering; qv; ROSA62; SpbIV; spectrin beta 4; spectrin beta chain, brain 3; spectrin beta chain, non-erythrocytic 4; spectrin beta, non-erythrocytic 4; spectrin, beta, non-erythrocytic 4; spectrin, non-erythroid beta chain 3; SPNB4; SPTBN3; SPTBN4 | |
| SPTBN4 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title