Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Speedglas™ Fabric Head Cover
Shop All 3M Products
Click to view available options
Size:
Large
Standard
Description
- A variety of accessories are available for a Speedglas welding helmet to help maximize protection and comfort
- For maximum safety and comfort, make it a habit to check equipment regularly
- Replace worn out or damaged parts.
- Large head coverage option is for use with the complete helmet assembly to help protect the user from welding sparks and spatter on the top portion of the head.
Specifications
Specifications
| Construction | Flame retardant |
| Color | Black |
| For Use With (Equipment) | Speedglas Heavy-duty Welding Helmet G5-01 |
| For Use With (Application) | Grinding, Welding, Metal Processing, Metal Repair |
| Model | 46-0700-80 |
| Product Type | Headcover |
| Quantity | 1 Ea. |
| Size | Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction