Learn More
3M™ Speedglas™ Welding Helmet Magnification Plate
Catalog No. 19070954 Shop All 3M Products
Click to view available options
Product Type:
Magnifying Lens 2.0X
Magnifying Lens 3.0X
The magnifier is used for magnifying objects while inserted into the Speedglas™ 9100 ADF on the inside slot.
  • Magnification Plate
  • Compatible 9100 Series ADFs
  • 3M Catalog# 06-0500-57 , 3M Catalog# 06-0500-59
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010
For Use With (Equipment) Speedglas 9100 ADF
For Use With (Application) Welding, Sanding, Grinding, Facility Maintenance, Machine Operations, Assembly
Lens Tint None
Model 06-0500-57
Product Type Magnifying Lens 2.0X
Quantity 1 Ea.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.