Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Speedy/Ringo Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB1002521F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002521F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002521F Supplier Novus Biologicals Supplier No. NB1002521F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Speedy/Ringo Polyclonal specifically detects Speedy/Ringo in Human, Mouse, Rat, Sheep samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Speedy/Ringo
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q5MJ70
Gene Alias hSpy/Ringo A, MGC110856, Rapid inducer of G2/M progression in oocytes A, RINGO A, Ringo3, SPDY1, speedy homolog 1 (Drosophila), speedy homolog A (Xenopus laevis), speedy protein A, speedy-1, Spy1, SPY1MGC57218
Gene Symbols SPDYA
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human Speedy/Ringo (within residues 200-286). This immunogen is conserved in both isoforms 1 and 2. [Swiss-Prot Q5MJ69]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 245711
Test Specificity Mouse. Sheep reactivity reported in scientific literature (PMID: 19187565). Predicted to react with human based on 100% sequence homology. Human reactivity reported in scientific literature (PMID: 24037419).
Target Species Human, Mouse, Rat, Sheep
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.