Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Speedy/Ringo Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB339784 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339784 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339784 Supplier Novus Biologicals Supplier No. NB1002521MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Speedy/Ringo Polyclonal specifically detects Speedy/Ringo in Human, Mouse, Rat, Sheep samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Speedy/Ringo
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias hSpy/Ringo A, MGC110856, Rapid inducer of G2/M progression in oocytes A, RINGO A, Ringo3, SPDY1, speedy homolog 1 (Drosophila), speedy homolog A (Xenopus laevis), speedy protein A, speedy-1, Spy1, SPY1MGC57218
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human Speedy/Ringo (within residues 200-286). This immunogen is conserved in both isoforms 1 and 2. [Swiss-Prot Q5MJ69]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 245711
Target Species Human, Mouse, Rat, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.