Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEN Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291734 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291734 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291734 Supplier Novus Biologicals Supplier No. NB10058799APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPEN Polyclonal antibody specifically detects SPEN in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Proximity Ligation Assay, ChIP assay (ChIP)

Specifications

Antigen SPEN
Applications Western Blot, Immunoprecipitation, Proximity Ligation Assay, ChIP Assay
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation, Proximity Ligation Assay, Chromatin Immunoprecipitation (ChIP)
Formulation PBS
Gene Alias HIAA0929, KIAA0929nuclear receptor transcription cofactor, MINTMsx2 interacting nuclear target (MINT) homolog, RBM15C, SHARPmsx2-interacting protein, SMART/HDAC1 associated repressor protein, SMART/HDAC1-associated repressor protein, SPEN homolog, spen homolog, transcriptional regulator (Drosophila)
Host Species Rabbit
Immunogen Maps to a region between residue 3375 and 3425 of human SMART/HDAC1 associated repressor protein (spen homolog, transcriptional regulator) using the numbering given in entry NP_055816.2 (GeneID 23013).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23013
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.