Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEN Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291736 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291736 Supplier Novus Biologicals Supplier No. NB10058798PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPEN Polyclonal antibody specifically detects SPEN in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen SPEN
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias HIAA0929, KIAA0929nuclear receptor transcription cofactor, MINTMsx2 interacting nuclear target (MINT) homolog, RBM15C, SHARPmsx2-interacting protein, SMART/HDAC1 associated repressor protein, SMART/HDAC1-associated repressor protein, SPEN homolog, spen homolog, transcriptional regulator (Drosophila)
Host Species Rabbit
Immunogen Maps to a region between residue 1350 and 1400 of human SMART/HDAC1 associated repressor protein (spen homolog, transcriptional regulator) using the numbering given in entry NP_055816.2 (GeneID 23013).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23013
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.