Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermidine Antibody, Novus Biologicals™
SDP

Catalog No. NB1001847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001847 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001847 Supplier Novus Biologicals Supplier No. NB1001847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Spermidine Polyclonal specifically detects Spermidine in Human, Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Spermidine
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1200-1:5000, Immunohistochemistry 1:50 - 1:500, Immunohistochemistry-Paraffin 1:50-1:500
Formulation Antiserum with No Preservative
Gene Alias 1,5,10-Triazadecane, 124-20-9, 4-Azaoctamethylenediamine, 4-Azaoctane-1,8-diamine, N-(3-aminopropyl)butane-1,4-diamine, N1-(3-Aminopropyl)butane-1,4-diamine, Spermidin
Host Species Rabbit
Immunogen Spermidine conjugated to KLH.
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with spermidine and cross reacts with spermine.
Reconstitution Reconstitute with 0.2 ml distilled water.
Target Species Human, Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.