Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Grinding Balls for High-Energy Mixer/Mill™ Grinding Vial Sets

Catalog No. p-4878650
Click to view available options
Material:
Hardened Steel
Stainless Steel

Crush, grind, and mill various materials with these Grinding Balls for High-Energy Mixer/Mill™ Grinding Vial Sets.

  • Select from 10 different grinding ball materials to suit your application.
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.

Specifications

Product Type Grinding Balls
Composition Fe, Cr, Si, Mn, C
Diameter (English) 0.5 in., 0.25 in.
Diameter (Metric) 1.27 cm, 0.63 cm
For Use With (Application) Ideal for General Purpose Grinding
For Use With (Equipment) Vial Kit of 8000 D and 8000 M Mixer or Mill
Hardness 5.5 to 6
Material Hardened Steel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.