Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPFH2 Antibody (SR1636), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32243950UL
Description
SPFH2 Monoclonal antibody specifically detects SPFH2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
SPFH2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1636 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
C8orf2SPFH domain-containing protein 2, Endoplasmic reticulum lipid raft-associated protein 2, ER lipid raft associated 2, Erlin-2, MGC87072, NET32, SPFH domain family, member 2, SPFH2chromosome 8 open reading frame 2, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2 | |
A synthesized peptide derived from human SPFH2 (Uniprot #: O94905) | |
50 μg | |
Alzheimers Research, Neurodegeneration, Neuroscience, Signal Transduction | |
11160 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction